missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD170 (Siglec F) Monoclonal Antibody (1RNM44N), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™

Product Code. 30055795
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30055795 25 μg 25µg
30056121 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30055795 Supplier Invitrogen™ Supplier No. M042T02B08A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

SIGLEC5 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. This protein binds equally to alpha2,3-linked and alpha2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. This molecule is expressed by monocytic/myeloid lineage cells. SIGLEC5 is found at high levels in peripheral blood leukocytes, spleen, bone marrow and at lower levels in lymph node, lung, appendix, placenta, pancreas and thymus. It is expressed by monocytes and neutrophils but absent from leukemic cell lines representing early stages of myelomonocytic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD170 (Siglec F)
Applications Flow Cytometry
Classification Monoclonal
Clone 1RNM44N
Concentration 0.1 mg/mL
Conjugate NovaFluor Blue 660-120S
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Siglecf
Gene Accession No. Q920G3
Gene Alias CD170; CD33 antigen-like 2; CD33L2; mSiglec-F; OB-binding protein 2; OBBP2; OB-BP2; Obesity-binding protein 2; sialic acid binding Ig like lectin 5; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin F; sialic acid-binding Ig-like lectin 5; Sialic acid-binding Ig-like lectin F; sialic acid-binding immunoglobulin-like lectin 5; sialic acid-binding lectin Siglec-F; Siglec; SIGLEC5; siglec-5; Siglecf; Siglec-F
Gene Symbols Siglecf
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233186
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.