missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD170 (Siglec F) Monoclonal Antibody (1RNM44N), NovaFluor™ Red 685, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

178.00 € - 344.00 €

Specifications

Antigen CD170 (Siglec F)
Clone 1RNM44N
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30054846
View Documents
Invitrogen™
M042T03R02-A
100 μg
344.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055083
View Documents
Invitrogen™
M042T02R02-A
25 μg
178.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD170 (Siglec F) Monoclonal antibody specifically detects CD170 (Siglec F) in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD170 (Siglec F)
0.1 mg/mL
Monoclonal
Rat
Mouse
Q920G3
233186
Primary
4°C, store in dark, DO NOT FREEZE!
1RNM44N
Flow Cytometry
NovaFluor Red 685
RUO
PBS with 0.09% sodium azide; pH 7.2
CD170; CD33 antigen-like 2; CD33L2; mSiglec-F; OB-binding protein 2; OBBP2; OB-BP2; Obesity-binding protein 2; sialic acid binding Ig like lectin 5; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin F; sialic acid-binding Ig-like lectin 5; Sialic acid-binding Ig-like lectin F; sialic acid-binding immunoglobulin-like lectin 5; sialic acid-binding lectin Siglec-F; Siglec; SIGLEC5; siglec-5; Siglecf; Siglec-F
IgG2a κ
Affinity chromatography
Siglecf
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.