missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD181 (CXCR1) Monoclonal Antibody (eBio8F1-1-4 (8F1-1-4)), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
156.00 € - 305.00 €
Specifications
| Antigen | CD181 (CXCR1) |
|---|---|
| Clone | eBio8F1-1-4 (8F1-1-4) |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CXCR1 is a member of the G-protein-coupled receptor family. This protein is a receptor for interleukin 8 (IL8). It binds to IL8 with high affinity, and transduces the signal through a G-protein activated second messenger system. Knockout studies in mice suggested that this protein inhibits embryonic oligodendrocyte precursor migration in developing spinal cord. This gene, IL8RB, a gene encoding another high affinity IL8 receptor, as well as IL8RBP, a pseudogene of IL8RB, form a gene cluster in a region mapped to chromosome 2q33-q36.Specifications
| CD181 (CXCR1) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| 3577 | |
| IgG2b κ | |
| Affinity chromatography | |
| CXCR1 |
| eBio8F1-1-4 (8F1-1-4) | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| P25024 | |
| CXCR1 | |
| Primary | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title