missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (4G7-2E3R), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4867RCL1
This item is not returnable.
View return policy
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry
Specifications
| CD19 | |
| Monoclonal | |
| CoraFluor 1 | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Human CCL cells, | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer Stem Cells, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 930 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 4G7-2E3R | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction