missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD19 Monoclonal Antibody (HIB19), NovaFluor™ Blue 610-30S, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

192.00 € - 421.00 €

Specifications

Antigen CD19
Clone HIB19
Concentration 1.0 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055775
View Documents
Invitrogen™
H004T03B05-A
100 Tests
421.00 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30056199
View Documents
Invitrogen™
H004T02B05-A
25 Tests
192.00 €
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD19
1.0 μg/Test
Monoclonal
Mouse
Human
P15391
930
Primary
4°C, store in dark, DO NOT FREEZE!
HIB19
Flow Cytometry
NovaFluor Blue 610-30S
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12
IgG1 κ
Affinity chromatography
Cd19
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.