missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD19 Monoclonal Antibody (HIB19), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
192.00 € - 421.00 €
Specifications
| Antigen | CD19 |
|---|---|
| Clone | HIB19 |
| Concentration | 1.0 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055239
|
Invitrogen™
H004T03B06-A |
100 Tests |
421.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055746
|
Invitrogen™
H004T02B06-A |
25 Tests |
192.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow CytometrySpecifications
| CD19 | |
| 1.0 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P15391 | |
| 930 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| HIB19 | |
| Flow Cytometry | |
| NovaFluor Blue 610-70S | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
| IgG1 κ | |
| Affinity chromatography | |
| Cd19 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title