Learn More
CD19 Monoclonal Antibody (SJ25C1), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 13-0198-41
Description
Description: The SJ25C1 monoclonal antibody reacts with human CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express this molecule. CD19, along with CD21, CD81, Leu13, and MHC class II, form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux, and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes. Applications Reported: This SJ25C1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SJ25C1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry
Specifications
| CD19 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| P15391 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| SJ25C1 | |
| Biotin | |
| Cd19 | |
| AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
| Affinity chromatography | |
| RUO | |
| 930 | |
| 4° C, do not freeze | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.