missing translation for 'onlineSavingsMsg'
Learn More

CD19 Rat, PerCP-Cy5.5, Clone: 1D3, enQuireBio™

Product Code. 15911768
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15911768 100 μg 100µg
15921768 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15911768 Supplier EnquireBio Supplier No. QAB28PCP55100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Specifications

Antigen CD19
Applications Flow Cytometry
Classification Monoclonal
Clone 1D3
Concentration 0.5mg/mL
Conjugate PerCP-Cyanine5.5
Formulation 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Gene Accession No. NP_033974
Gene Symbols Cd19
Host Species Rat
Purification Method Protein A/G Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12478
Content And Storage Store at 2-4°C.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.