Learn More
Invitrogen™ CD199 (CCR9) Recombinant Mouse Monoclonal Antibody (BBC3M4)
Mouse Recombinant Monoclonal Antibody
199.00 € - 349.00 €
Specifications
| Antigen | CD199 (CCR9) |
|---|---|
| Clone | BBC3M4 |
| Concentration | 1.0 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | Flow Cytometry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30565063
|
Invitrogen™
740187M |
100 μL |
349.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30565024
|
Invitrogen™
740187MXS |
20 μL |
199.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant mouse monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
The protein encoded by this gene is a member of the beta chemokine receptor family. It is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are key regulators of the thymocytes migration and maturation in normal and inflammation conditions. The specific ligand of this receptor is CCL25. It has been found that this gene is differentially expressed by T lymphocytes of small intestine and colon, suggested a role in the thymocytes recruitment and development that may permit functional specialization of immune responses in different segment of the gastrointestinal tract. This gene is mapped to the chemokine receptor gene cluster region. Two alternatively spliced transcript variants have been described.Specifications
| CD199 (CCR9) | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P51686 | |
| 10803 | |
| Primary | |
| Protein A/G |
| BBC3M4 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| Ccr9 | |
| A130091K22Rik; C-C chemokine receptor type 9; C-C CKR-9; C-C motif chemokine receptor 9; CC-CKR-9; Ccr9; CCR-9; CDw199; CDw199 antigen; chemokine (C-C motif) receptor 9; chemokine C-C receptor 10; Cmkbr10; G protein-coupled receptor 28; GPR28; GPR-9-6; G-protein coupled receptor 28 | |
| Ccr9 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.