missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1c/BDCA-1 Antibody (SR1634), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22372-50ul
This item is not returnable.
View return policy
Description
CD1c/BDCA-1 Monoclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western Blot
Specifications
| CD1c/BDCA-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1634 | |
| Western Blot 1:500-1:2000 | |
| BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
| A synthesized peptide derived from human CD1c/BDCA-1 (Uniprot #: P29017) | |
| 50 μg | |
| Cancer, Immunology | |
| 911 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction