missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD1c Monoclonal Antibody (L161), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
185.00 € - 392.00 €
Specifications
| Antigen | CD1c |
|---|---|
| Clone | L161 |
| Concentration | 0.1 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055040
|
Invitrogen™
H046T03R06-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055742
|
Invitrogen™
H046T02R06-A |
25 Tests |
185.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD1c Monoclonal antibody specifically detects CD1c in Human samples. It is validated for Flow CytometrySpecifications
| CD1c | |
| 0.1 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P29017 | |
| 911 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| L161 | |
| Flow Cytometry | |
| NovaFluor Red 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| BDCA1; canCD1c; CD1; CD1A; CD1C; CD1C antigen, c polypeptide; CD1c molecule; cortical thymocyte antigen CD1C; differentiation antigen CD1-alpha-3; R7; RP11-101J8.3; T-cell surface glycoprotein CD1c | |
| IgG1 κ | |
| Affinity chromatography | |
| CD1C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title