missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Antibody (1B1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
438.00 € - 593.00 €
Specifications
| Antigen | CD1d |
|---|---|
| Clone | 1B1 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18720084
|
Novus Biologicals
NBP1-43461-0.1MG |
0.1 mg |
438.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18206721
|
Novus Biologicals
NBP1-43461-0.5MG |
0.5 mg |
593.00 €
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD1d Monoclonal specifically detects CD1d in Mouse, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation.Specifications
| CD1d | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Mouse | |
| antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D | |
| CD1D | |
| IgG2b κ | |
| Affinity Purified |
| 1B1 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Rat | |
| B Cell Development and Differentiation Markers, Myeloid derived Suppressor Cell | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 912 | |
| The immunogen for this antibody was CD1d. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title