missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD1d Antibody (3A6G3), Novus Biologicals™
Mouse Monoclonal Antibody
287.00 € - 511.00 €
Specifications
| Antigen | CD1d |
|---|---|
| Clone | 3A6G3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30031964
|
Novus Biologicals™
NBP3-27170-0.1ml |
0.1 mL |
511.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30038490
|
Novus Biologicals™
NBP3-27170-0.025ml |
0.025 mL |
287.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD1d Monoclonal antibody specifically detects CD1d in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| CD1d | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, B Cell Development and Differentiation Markers, Cellular Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Neuroscience | |
| PBS | |
| 912 | |
| IgG2b | |
| Protein G purified |
| 3A6G3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D | |
| Purified recombinant fragment of human CD1d (AA: extra 20-301) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title