missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (rLFA2/8516), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20398-100ug
This item is not returnable.
View return policy
Description
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Recombinant fragment of human CD2 protein (exact sequence is proprietary) | |
| 100 μg | |
| Adaptive Immunity, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells | |
| 914 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rLFA2/8516 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction