missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Mouse, V450, Clone: RPA-2.10, enQuireBio™
Mouse Monoclonal Antibody
203.00 € - 2225.00 €
Specifications
Antigen | CD2 |
---|---|
Clone | RPA-2.10 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951468
|
EnquireBio
QAB1-V450-25TESTS |
25 Tests |
203.00 €
25 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941468
|
EnquireBio
QAB1-V450-100TESTS |
100 Tests |
505.00 €
100 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961468
|
EnquireBio
QAB1-V450-500TESTS |
500 Tests |
2225.00 €
500 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD2 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
914 | |
IgG1 κ | |
Protein A/G Purified |
RPA-2.10 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
NP_001758 | |
CD2 | |
Primary | |
Store at 2-4°C. |