missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2, Rat anti-Human, Clone: YTH 655, Novus Biologicals™
Rat Monoclonal Antibody
280.00 € - 583.00 €
Specifications
| Antigen | CD2 |
|---|---|
| Clone | YTH 655 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Cell depletion |
| Applications | Flow Cytometry, Depletion Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246559
|
Novus Biologicals
NBP2-81042-0.025MG |
0.025 mg |
280.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18275609
|
Novus Biologicals
NBP2-81042-0.2MG |
0.2 mg |
583.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Cell depletion.Specifications
| CD2 | |
| 1 mg/mL | |
| Flow Cytometry, Depletion Assay | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Apoptosis, Immunology | |
| PBS | |
| 914 | |
| Recombinant monoclonal antibody to CD2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| YTH 655 | |
| Flow Cytometry, Cell depletion | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| CD2 | |
| IgG2b κ | |
| Protein A purified | |
| This antibody specifically recognizes human CD2, a cell adhesion molecule found on the surface of T cells and natural killer (NK) cells. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title