missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (109-3C2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
552.00 € - 597.00 €
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 109-3C2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699065
|
Novus Biologicals
NBP2-47841B-100ul |
100 μg |
597.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672606
|
Novus Biologicals
NBP2-47841B |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for ELISASpecifications
| CD20 | |
| ELISA | |
| Biotin | |
| Mouse | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| PBS | |
| 931 | |
| IgG3 κ | |
| Protein A or G purified |
| 109-3C2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Stimulated human leukocytes | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title