missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (10F381 (rituximab)) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
281.00 € - 569.00 €
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 10F381 (rituximab) |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668618
|
Novus Biologicals
NBP2-75897 |
0.2 mg |
569.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638897
|
Novus Biologicals
NBP2-75897SS |
0.025 mg |
281.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD20 Monoclonal antibody specifically detects CD20 in Human, Cynomolgus Monkey, Macaque samples. It is validated for Flow CytometrySpecifications
| CD20 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Cynomolgus Monkey, Macaque | |
| B1, biosimilar, biosimilars, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Human lymphoblastoid cell line SB. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| 10F381 (rituximab) | |
| Flow Cytometry | |
| Unconjugated | |
| Human | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| PBS | |
| 931 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title