missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (IGEL/1497R), Novus Biologicals™
Rabbit Monoclonal Antibody
339.00 € - 553.00 €
Specifications
| Antigen | CD20 |
|---|---|
| Clone | IGEL/1497R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681538
|
Novus Biologicals
NBP2-49872SS |
0.02 mg |
339.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645477
|
Novus Biologicals
NBP2-49872-0.1mg |
0.1 mg |
553.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CD20 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 10 mM PBS with 0.05% BSA | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Recombinant human full-length CD20 protein (Uniprot: P11836) | |
| Primary | |
| Store at 4°C. |
| IGEL/1497R | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P11836, P11836 | |
| 931 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title