Learn More
CD20 Mouse anti-Human, PE, Clone: 2H7, BD
Mouse Monoclonal Antibody
74.20 € - 166.92 €
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 2H7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15840639
|
BD Biosciences
560961 |
25 Tests |
78.15 € 74.20 € / Each Save 3.95 € 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15898308
|
BD Biosciences
555623 |
100 Tests |
174.00 € 166.92 € / Each Save 7.08 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 2H7 monoclonal antibody specifically binds to CD20 that is encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells and follicular dendritic cells but not on plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Specifications
| CD20 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG2b κ | |
| Affinity Purified |
| 2H7 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human 6.16c1.3 B cell line | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.