missing translation for 'onlineSavingsMsg'
Learn More

CD20 Mouse, Qfluor 710, Clone: 2H7, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB31-QF710-100Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961838

  • 510.00 € / 100 tests
Estimated Shipment: 20-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD20
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
MS4A1
Protein A/G Purified
RUO
931
IgG2b κ
Flow Cytometry
2H7
Qfluor 710
NP_690605
Mouse
100 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers