missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 / MS4A1 Rabbit, Clone: IGEL/1497R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | CD20 / MS4A1 |
|---|---|
| Clone | IGEL/1497R |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15921198
|
EnquireBio
931-RBM7-P0 |
20 μg | |||||||
|
15931198
|
EnquireBio
931-RBM7-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD20 / MS4A1 | |
| 200μg/mL | |
| Monoclonal | |
| Rabbit | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 931 | |
| Recombinant human full-length MS4A1 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 33-37kDa |
| IGEL/1497R | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_690605 | |
| MS4A1 | |
| IgG κ | |
| Protein A/G Purified | |
| Predominantly cell surface with some cytoplasmic | |
| Daudi, Raji, and U266, and human lymphocytes. Lymph nodes and tonsils. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title