missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD200 Monoclonal Antibody (OX104), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD200 |
|---|---|
| Clone | OX104 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055488
|
Invitrogen™
H038T03R04-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054928
|
Invitrogen™
H038T02R04-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow CytometrySpecifications
| CD200 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P41217 | |
| 4345 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| OX104 | |
| Flow Cytometry | |
| NovaFluor Red 710 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
| IgG1 κ | |
| Affinity chromatography | |
| Cd200 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title