missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06398-100ul
This item is not returnable.
View return policy
Description
CD200R1L Polyclonal antibody specifically detects CD200R1L in Human samples. It is validated for ELISA
Specifications
| CD200R1L | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa | |
| Produced in rabbits immunized with purified, recombinant Human CD200R1L (Accession#: AAT00538.1; Met1-Leu239) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 344807 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction