Learn More
Invitrogen™ CD203c Recombinant Rabbit Monoclonal Antibody (8F10L7)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558179
Description
Immunogen sequence: SHAEEVSKFS VCGFANPLPT ESLDCFCPHL QNSTQLEQVN QMLNLTQEEI TATVKVNLPF GRPRVLQKNV DHCLLYHREY VSGFGKAMRM PMWSSYTVPQ LGDTSPLPPT VPDCLRADVR VPPSESQKCS FYLADKNITH GFLYPPASNR TSDSQYDALI TSNLVPMYEE FRKMWDYFHS VLLIKHATER NGVNVVSGPI FDYNYDGHFD APDEITKHLA NTDVPIPTHY FVVLTSCKNK SHTPENCPGW LDVLPFIIPH RPTNVESCPE GKPEALWVEE RFTAHIARVR DVELLTGLDF YQDKVQPVSE ILQLKTYLPT FETTI.
The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq, Jul 2008].
Specifications
| CD203c | |
| Recombinant Monoclonal | |
| 0.55 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O14638 | |
| ENPP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 551-875 of human ENPP3/CD203c (NP_005012.2). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 8F10L7 | |
| Unconjugated | |
| ENPP3 | |
| AI876438; alkaline phosphodiesterase; Alkaline phosphodiesterase I; B10; CD203c; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); ectonucleotide pyrophosphatase/phosphodiesterase 3; ectonucleotide pyrophosphatase/phosphodiesterase family member 3; E-NPP 3; ENPP3; gp130RB13-6; Npp3; NPPase; Nucleotide diphosphatase; Nucleotide pyrophosphatase; PD-Ibeta; Pdnp3; Phosphodiesterase I beta; phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta; RB13-6; RB13-6 antigen | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5169 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.