missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD205 Monoclonal Antibody (205yekta), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
185.00 € - 348.00 €
Specifications
| Antigen | CD205 |
|---|---|
| Clone | 205yekta |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056087
|
Invitrogen™
M051T03R06-A |
100 μg |
348.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055189
|
Invitrogen™
M051T02R06-A |
25 μg |
185.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD205 Monoclonal antibody specifically detects CD205 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD205 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q60767 | |
| 17076 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 205yekta | |
| Flow Cytometry | |
| NovaFluor Red 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CA-IX; CD205; CLEC13B; C-type lectin domain family 13 member B; DCL-1 fusion; DEC205; DEC-205; DEC-205/CD205 protein; gp200; gp200-MR6; group VI C-type animal lectin; LY75; ly-75; Lymphocyte antigen 75; membrane antigen MN; P54/58N; pMW1; RGD1565962; RP23-10L17.4 | |
| IgG2a κ | |
| Affinity chromatography | |
| LY75 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title