missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Antibody (BB6-11C9.6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-28245
This item is not returnable.
View return policy
Description
CD21 Monoclonal specifically detects CD21 in Mouse, Porcine samples. It is validated for Flow Cytometry.
Specifications
| CD21 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| CR2 | |
| Porcine CD21, Mr 160 kDa | |
| 0.25 mg | |
| Dendritic Cell Markers, Immunology | |
| 1380 | |
| Mouse, Pig | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| BB6-11C9.6 | |
| Unconjugated | |
| C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Porcine CD21, Mr 160 kDa | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction