missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD21 Recombinant Rabbit Monoclonal Antibody (257), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
109.00 € - 326.00 €
Specifications
| Antigen | CD21 |
|---|---|
| Clone | 257 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019279
![]() |
Invitrogen™
MA552250 |
100 Tests |
326.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019492
|
Invitrogen™
MA552251 |
25 Tests |
109.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD21 Recombinant Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Flow CytometrySpecifications
| CD21 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.09% sodium azide | |
| C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9 | |
| Recombinant Human CD21 Protein (Ile 21-Arg971). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CR2 |
| 257 | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| Human | |
| P20023 | |
| 1380 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title