missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD229 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148544
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
CD229 (Ly9) is a cell surface receptor of the CD150 family, which includes also e.g. CD48 and CD224. Receptors of this family regulate cytokine production and cytotoxicity of lymphocytes and NK cells. High levels of CD229 are found on T and B cells, where its expression increases during their maturation. It is absent on granulocytes, bone marrow-derived dendritic cells, platelets and erythrocytes. CD229 has been also reported on mouse monocytes and NK cells. CD229 interacts homophilically through its N-terminal domain and localizes to the contact site between T cells and antigen presenting B cells during antigen-dependent immune synapse formation.
Specifications
| CD229 | |
| Polyclonal | |
| Unconjugated | |
| Ly9 | |
| AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; LY9; Ly-9; Lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 4063 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9HBG7 | |
| Ly9 | |
| Mouse myeloma cell line NS0-derived recombinant human CD229/SLAMF3, Lys48-Lys454. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction