missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (249936), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1898C
This item is not returnable.
View return policy
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal antibody specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CD229/SLAMF3/Lymphocyte Antigen 9 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 249936 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
| Mouse myeloma cell line NS0-derived recombinant human CD229/SLAMF3, Lys48-Lys454, Accession # Q9HBG7 | |
| 0.1 mL | |
| Immunology | |
| 4063 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction