Learn More
CD23/Fc epsilon RII Antibody (EBVCS2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-43433-0.1MG
 View more versions of this product
Product Code. 18272853
  • 474.00 € / 0.10mg
Description
CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD23/Fc epsilon RII
Monoclonal
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
FCER2
The immunogen for this antibody was CD23.
0.1 mg
Adaptive Immunity, Cancer, Immunology
2208
Store at 4C. Do not freeze.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
EBVCS2
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Mouse
Affinity Purified
RUO
Primary
Human
IgG1