missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23 Rat anti-Mouse, Oligo, Clone: B3B4, BD™ AbSeq ™
Rat Monoclonal Antibody
Brand: BD Biosciences 940161
Additional Details : Weight : 0.00010kg
Description
CD23 Monoclonal antibody specifically detects CD23 in Mouse samples. It is validated for Single Cell 3' SequencingSpecifications
CD23 | |
Monoclonal | |
Oligo | |
FcεRII; Fc-epsilon-RII; Fcer2a; Ly-42; Low-affinity IgE receptor; Fcer2 | |
FcεR isolated from the mouse B hybridoma line O1.2B2 | |
RUO | |
Mouse | |
IgG2a κ |
Single Cell 3' Sequencing | |
Clone B3B4 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |