Learn More
CD235ab (Glycophorin A/B) Mouse, anti-Human, Unconjugated, Clone: HIR2 (GA-R2), STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60111
Encompass_Preferred
 View more versions of this product
Product Code. 17119101
  • 194.40 € / 100µg
Description
  • The HIR2 (GA-R2) antibody recognizes an epitope common to the N-terminal region of human CD235a (glycophorin A) and CD235b (glycophorin B), homologous type I sialoglycoproteins of the erythrocyte (red blood cell) membrane that bear the antigenic determinants for the MN and Ss blood groups
  • CD235ab is expressed on early- and late-stage erythroblasts, mature erythrocytes, and erythroid cell lines such as K562 and HEL, but not on other cell types
  • CD235a is abundantly expressed whereas CD235b is a relatively minor membrane component
  • The proteins are believed to provide a large mucin-like surface to erythrocytes that acts to minimize aggregation in the circulation
  • The HIR2 antibody binds with higher affinity to CD235a than CD235b, and agglutinates untreated erythrocytes
Specifications
Specifications
CD235ab (Glycophorin A/B)
Monoclonal
Unconjugated
100 μg
Human
Red Blood Cells
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
HIR2 (GA-R2)
Mouse
Primary
IgG2b κ