missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
203.00 € - 552.00 €
Specifications
| Antigen | CD24 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18208034
|
Novus Biologicals
NBP1-46390SS |
0.025 mL |
203.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18765113
|
Novus Biologicals
NBP1-46390 |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD24 Polyclonal specifically detects CD24 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CD24 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P25063 | |
| 100133941 | |
| A synthetic peptide from aa region 30-70 of human mature Signal transducer CD24 conjugated to blue carrier protein was used as the antigen. | |
| Primary | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Specific for CD24. |
| Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| Lyophilized from whole antisera with No Preservative | |
| CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
| CD24 | |
| IgG | |
| Unpurified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title