missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD24 |
|---|---|
| Clone | eBioSN3 (SN3 A5-2H10) |
| Concentration | 0.4 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055357
|
Invitrogen™
H064T03B01-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056109
|
Invitrogen™
H064T02B01-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Flow CytometrySpecifica
| CD24 | |
| 0.4 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P25063 | |
| 100133941 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBioSN3 (SN3 A5-2H10) | |
| Flow Cytometry | |
| NovaFluor Blue 510 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen | |
| IgG1 κ | |
| Affinity chromatography | |
| Cd24 |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto