missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), NovaFluor™ Yellow 570, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

178.00 € - 392.00 €

Specifications

Antigen CD24
Clone eBioSN3 (SN3 A5-2H10)
Concentration 0.2 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30056188
View Documents
Invitrogen™
H064T03Y01-A
100 Tests
392.00 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055882
View Documents
Invitrogen™
H064T02Y01-A
25 Tests
178.00 €
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD24
0.2 μg/Test
Monoclonal
Mouse
Human
P25063
100133941
Primary
4°C, store in dark, DO NOT FREEZE!
eBioSN3 (SN3 A5-2H10)
Flow Cytometry
NovaFluor Yellow 570
RUO
PBS with 0.09% sodium azide; pH 7.2
CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen
IgG1 κ
Affinity chromatography
Cd24
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.