Learn More
CD24 Mouse anti-Human, FITC, Clone: ML5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560992
Description
The ML5 monoclonal antibody specifically binds to CD24. CD24 is a 35-45 kDa two-chain glycoprotein expressed on the surface of B cells, granulocytes and most B-cell lines. CD24 may play a role in the regulation of B-cell proliferation and maturation.
Specifications
| CD24 | |
| Monoclonal | |
| FITC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| ML5 | |
| Heat Stable Antigen Homologue (HSA); Ba-1; CD24A | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.