missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Rat, APC, Clone: M1/69, enQuireBio™
Rat Monoclonal Antibody
155.00 € - 469.00 €
Specifications
Antigen | CD24 |
---|---|
Clone | M1/69 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991848
|
EnquireBio
QAB33-APC-25UG |
25 μg |
155.00 €
25µg |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981848
|
EnquireBio
QAB33-APC-100UG |
100 μg |
469.00 €
100µg |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD24 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
12484 | |
IgG2b κ | |
Protein A/G Purified |
M1/69 | |
Flow Cytometry | |
APC | |
RUO | |
NP_033976 | |
Cd24a | |
Primary | |
Store at 2-4°C. |