missing translation for 'onlineSavingsMsg'
Learn More

CD247 Hamster anti-Human, FITC, Clone: H146-968, Invitrogen™

Product Code. 15285834
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15285834 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15285834 Supplier Invitrogen Supplier No. A15754

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hamster Monoclonal Antibody

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD247
Applications Flow Cytometry
Classification Monoclonal
Clone H146-968
Conjugate FITC
Formulation PBS, pH 7.4, with 0.2% BSA and 15mM sodium azide
Gene Alias RP11-104L21.1, CD3-ZETA, CD3H, CD3Q, CD3Z, T3Z, TCRZ
Host Species Hamster
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 919
Target Species Human
Content And Storage Store at 2 - 8 °C.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.