missing translation for 'onlineSavingsMsg'
Learn More

CD25/ IL2RA (Activated Lymphocyte Marker) Mouse, Clone: IL2RA/2395, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 3559-MSM5-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960268

  • 462.00 € / 100µg
Estimated Shipment: 22-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD25/ IL2RA (Activated Lymphocyte Marker)
Cell Surface and Cytoplasmic
IL2RA/2395
Unconjugated
Mouse
55kDa
Protein A/G purified from Bioreactor concentrate.
RUO
3559
IgG2b
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinantfragment of human IL2RA protein (apprx. aa 42-183)
PHA-stimulated human lymphocytes. Human lymph nodeor tonsil
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers