missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD27 Monoclonal Antibody (O323), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD27 |
|---|---|
| Clone | O323 |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055462
|
Invitrogen™
H012T03R03-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055844
|
Invitrogen™
H012T02R03-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow CytometrySpecifications
| CD27 | |
| 0.8 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P26842 | |
| 939 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| O323 | |
| Flow Cytometry | |
| NovaFluor Red 700 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD27 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title