missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), NovaFluor™ Yellow 690, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00 € - 344.00 €
Specifications
| Antigen | CD274 (PD-L1, B7-H1) |
|---|---|
| Clone | MIH5 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056273
|
Invitrogen™
M036T03Y05-A |
100 μg |
344.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055550
|
Invitrogen™
M036T02Y05-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD274 (PD-L1, B7-H1) Monoclonal antibody specifically detects CD274 (PD-L1, B7-H1) in Mouse samples. It is validated for Flow CytometrySpecifications
| CD274 (PD-L1, B7-H1) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q9EP73 | |
| 60533 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| MIH5 | |
| Flow Cytometry | |
| NovaFluor Yellow 690 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
| IgG2a λ | |
| Affinity chromatography | |
| Cd274 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title