Learn More
CD275 (B7-H2) Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547177
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse B7-H2 and recombinant human B7-H1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD275 (B7-H2) Polyclonal antibody specifically detects CD275 (B7-H2) in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD275 (B7-H2) | |
Polyclonal | |
Unconjugated | |
ICOSLG | |
AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23308 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O75144 | |
ICOSLG | |
Mouse myeloma cell line NS0-derived recombinant human B7-H2 Asp19-Ser258 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |