missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD279 (PD-1) Monoclonal Antibody (J43), Brilliant Violet™ 786, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
171.00 € - 362.00 €
Specifications
| Antigen | CD279 (PD-1) |
|---|---|
| Clone | J43 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18486478
![]() |
Invitrogen™
417-9985-82 |
100 μg |
362.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18426508
|
Invitrogen™
417-9985-80 |
25 μg |
171.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow CytometrySpecifications
| CD279 (PD-1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| Mouse | |
| Q02242 | |
| 18566 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| J43 | |
| Flow Cytometry | |
| Brilliant Violet 786 | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
| IgG | |
| Affinity chromatography | |
| Pdcd1 |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title