missing translation for 'onlineSavingsMsg'
Learn More

CD279 (PD-1) Mouse anti-Human, BB515, Clone: EH12.1, BD Horizon™

Mouse Monoclonal Antibody

142.02 € - 205.44 €

Specifications

Antigen CD279 (PD-1)
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Blue 515 (BB515)
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15864609
View Documents
BD Biosciences
565936
25 Tests
ONLINE EXCLUSIVE PRICE
149.00 €
142.02 € / Each
Save 6.98 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15803689
View Documents
BD Biosciences
564494
50 Tests
ONLINE EXCLUSIVE PRICE
214.00 €
205.44 € / Each
Save 8.56 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometry.

CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

CD279 (PD-1)
Monoclonal
Purified
RUO
Human
hPD-1; PD1; PDCD1; Programmed cell death 1; SLEB2
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Flow Cytometry
Brilliant Blue 515 (BB515)
Mouse
Cell Biology
Aqueous buffered solution containing ≤0.09% sodium azide.
Human PD-1 Recombinant Protein
Primary
Vol. Per test: 5μL
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.