missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28, Rabbit anti-Mouse, Clone: D665, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81054-0.2MG
This item is not returnable.
View return policy
Beschreibung
CD28 Monoclonal specifically detects CD28 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Functional, Surface Plasmon Resonance, Block/Neutralize.
Spezifikation
| CD28 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Functional, Surface Plasmon Resonance, Block/Neutralize | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody binds to the laterally exposed C'D loop of CD28 and induces T cell proliferation without TCR ligation. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| Flow Cytometry, Immunohistochemistry, Functional Assay, Blocking Assay | |
| D665 | |
| Unconjugated | |
| PBS | |
| CD28 | |
| The original D665 antibody was generated by immunizing mice alternately with A20 cells expressing mCD28 i.p. and with recombinant mCD28Ig s.c. | |
| 0.2 mg | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 940 | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur