missing translation for 'onlineSavingsMsg'
Learn More

CD3 Antibody (SP7), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NB600-1441SS

 View more versions of this product

Product Code. 18798173

  • 262.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD3 Monoclonal antibody specifically detects CD3 in Human, Mouse, Porcine, Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Knockdown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD3
Monoclonal
Unconjugated
Tissue culture supernatant with 0.05% Sodium Azide
CD3
Synthetic peptide: KAKAKPVTRGAGA, corresponding to amino acids 156-168 of Human CD3 epsilon chain.
Tissue culture supernatant
RUO
Primary
The CD3 antigen is present on early thymocytes and mature T cells and is generally regarded as a pan-T cell marker. This antibody will help detect CD3 expression in normal and neoplastic tissues. This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cells in both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody is suitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
SP7
Flow Cytometry, Immunohistochemistry 1:25-1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:25-1:50, Immunohistochemistry-Frozen 1:10 - 1:500, KnockDown Validated
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Rabbit
23.1 kDa
0.1 mL
Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells
916
Human, Mouse, Pig, Canine
Supernatant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.