missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 gamma Antibody (CD3G/8794), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24120F
This item is not returnable.
View return policy
Description
CD3 gamma Monoclonal antibody specifically detects CD3 gamma in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD3 gamma | |
| Monoclonal | |
| FITC | |
| CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ79544, FLJ94613, MGC138597, T3G, T-cell antigen receptor complex, gamma subunit of T3, T-cell receptor T3 gamma chain, T-cell surface glycoprotein CD3 gamma chain | |
| Recombinant fragment (around aa1-182) of human CD3 gamma protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Innate Immunity | |
| 917 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin) | |
| CD3G/8794 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction