missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, PE-Cy5, Clone: HIT3A, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555341

 View more versions of this product

Product Code. 15858078

  • ONLINE EXCLUSIVE PRICE
    268.00 €
    257.53 € / Each
    Save 10.47 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The HIT3a monoclonal antibody specifically binds to the human CD3 ε-chain, a 20 kDa subunit of the CD3/T cell antigen receptor complex found on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody can be mitogenic for T lymphocytes. The CD3 complex plays a role in signal transduction during antigen recognition by the T cell receptor for antigen. HIT3a antibody does not stain intracellular CD3 unlike the other CD3-specific clone, UCHT1.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD3
Monoclonal
PE-Cyanine5
Mouse
100 Tests
Immunology
Human
IgG2a κ
Flow Cytometry
HIT3A
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.