missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, APC, Clone: SK7, enQuireBio™
Mouse Monoclonal Antibody
240.00 € - 2440.00 €
Specifications
Antigen | CD3 |
---|---|
Clone | SK7 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15921948
|
EnquireBio
QAB4-APC-25TESTS |
25 Tests |
240.00 €
25 tests |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911948
|
EnquireBio
QAB4-APC-100TESTS |
100 Tests |
535.00 €
100 tests |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931948
|
EnquireBio
QAB4-APC-500TESTS |
500 Tests |
2440.00 €
500 tests |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD3 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
916 | |
IgG1 κ | |
Protein A/G Purified |
SK7 | |
Flow Cytometry | |
APC | |
RUO | |
NP_000724 | |
CD3E | |
Primary | |
Store at 2-4°C. |